Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (mFluor™ UV375)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

104410X0 104410X1
AATB104410X0EA 2770 DKK
AATB104410X0 AATB104410X1
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (mFluor™ UV375)
Antistoffer
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Advarsel: For research use only.

Type: Primary
Antigen: CD44 antigen
Clonality: Monoclonal
Clone: IM7
Conjugation: mFluor™ UV375
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR