Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (mFluor™ Green 620)
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
104410U0 104410U1
AATB104410U0EA 2770 DKK
AATB104410U0 AATB104410U1
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (mFluor™ Green 620)
Antistoffer
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Advarsel: For research use only.
Type: Primary
Antigen: CD44 antigen
Clonality: Monoclonal
Clone: IM7
Conjugation: mFluor™ Green 620
Epitope:
Host: Rat
Isotype:
Reactivity: Human