Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (mFluor™ Green 620)
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
101920U0 101920U1
AATB101920U0EA 2770 DKK
AATB101920U0 AATB101920U1
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (mFluor™ Green 620)
Antistoffer
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Advarsel: For research use only.
Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: HIB19
Conjugation: mFluor™ Green 620
Epitope:
Host: Mouse
Isotype:
Reactivity: Human