Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 810)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

101920O0 101920O1
AATB101920O0EA 2770 DKK
AATB101920O0 AATB101920O1
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 810)
Antistoffer
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Advarsel: For research use only.

Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: HIB19
Conjugation: iFluor® 810
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR