Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 800)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

100410N0 100410N1
AATB100410N0EA 2770 DKK
AATB100410N0 AATB100410N1
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 800)
Antistoffer
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Advarsel: For research use only.

Type: Primary
Antigen: CD4 antigen
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: iFluor® 800
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR