Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 594)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

104410C0 104410C1
AATB104410C0EA 2770 DKK
AATB104410C0 AATB104410C1
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 594)
Antistoffer
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Advarsel: For research use only.

Type: Primary
Antigen: CD44 antigen
Clonality: Monoclonal
Clone: IM7
Conjugation: iFluor® 594
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR