Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 546)
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10041080 10041081
AATB10041080EA 2770 DKK
AATB10041080 AATB10041081
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 546)
Antistoffer
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Advarsel: For research use only.
Type: Primary
Antigen: CD4 antigen
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: iFluor® 546
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human