Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PacOrange)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

100321K0 100321K1
AATB100321K0EA 2770 DKK
AATB100321K0 AATB100321K1
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PacOrange)
Antistoffer
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kD member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependents).

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Advarsel: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: PacOrange
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR