Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (mFluor™ Violet 450)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

100520Z0 100520Z1
AATB100520Z0EA 2770 DKK
AATB100520Z0 AATB100520Z1
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (mFluor™ Violet 450)
Antistoffer
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Advarsel: For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: mFluor™ Violet 450
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR