Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 532)
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10052070 10052071
AATB10052070EA 2770 DKK
AATB10052070 AATB10052071
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 532)
Antistoffer
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Advarsel: For research use only.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: iFluor® 532
Epitope:
Host: Mouse
Isotype:
Reactivity: Human