Alcohol Plus Hand Sanitisers
Leverandør: Rubbermaid Commercial Products
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2162581 2146691 2146693 2146692 2146695
RUBB2162581EA 185 DKK
RUBB2162581 RUBB2146691 RUBB2146693 RUBB2146692 RUBB2146695
Alcohol Plus Hand Sanitisers
Hudpleje Sæber og huden rensemidler
Effective and fast-acting hand sanitiser is fully tested and proven to be bactericidal, virucidal, and yeasticidal and kills 99,99% of common germs.
  • Fast-acting formulation disinfects hands in 30 seconds
  • Contains 70% alcohol in line with WHO recommendations
  • Contains moisturising ingredients to help prevent skin dryness
Certificeringer: The Alcohol Plus formulation has been tested against the following standards: EN 1500, EN 1276, EN 13727 + A2, EN 14476 + A1, EN 14476 + A2, EN 1650 + A1, and EN 13624.