Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
100411U0 100411U1 100411U2
AATB100411U0EA 2380 DKK
AATB100411U0 AATB100411U1 AATB100411U2
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Antistoffer
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Advarsel: For research use only.
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human