Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 840)

Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

100210Q0 100210Q1
AATB100210Q0EA 2770 DKK
AATB100210Q0 AATB100210Q1
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 840)
Antistoffer
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Advarsel: For research use only.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: iFluor® 840
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR