Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 633)
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
100320E0 100320E1
AATB100320E0EA 2770 DKK
AATB100320E0 AATB100320E1
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 633)
Antistoffer
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kDa member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependent)s.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Advarsel: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: iFluor® 633
Epitope:
Host: Mouse
Isotype:
Reactivity: Human