Phenolphthalein 0,5% (w/v) in alcohol
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
CAS No.: 77-09-8
Molecular Formula: C20H14O4
Formula Weight: 318.33
Flash Point: 11°(52°F)
Physical Form: Liquid
Density: 0.918
Appearance: Colourless
Odour: Alcohol-like
UN No.: 1987
MDL No.: MFCD00005913
Formel:
C₂₀H₁₄O₄ Massefylde: 0,918 Flammepunkt: 11 °C (52 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005913 CAS-nummer: 77-09-8 EINECS: 201-004-7 UN nummer: 1987 ADR: 3,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...