Anti-Rabbit IgG (H+L) Donkey Polyclonal Antibody (CF®583R)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20895-50UL 20895-500UL
20895-50ULEA 765 DKK
20895-50UL 20895-500UL
Anti-Rabbit IgG (H+L) Donkey Polyclonal Antibody (CF®583R)
Antistoffer
This is a highly cross-adsorbed donkey anti-rabbit IgG (H+L) secondary antibody labeled with our bright and photostable CF® Dyes. To minimise cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, horse, human, mouse, rat, and sheep serum.

  • Highly cross-adsorbed for specific staining with minimal background
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues

Type: Secondary
Antigen: Rabbit IgG
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Donkey
Isotype: IgG (H+L)
Reactivity: Rabbit
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR