Methylrød, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

MQ200

1.06076.0025 1.06076.0100
1.06076.0025EA 611 DKK
1.06076.0025 1.06076.0100
Methylrød, Supelco®
Methylrød
Formel: C₁₅H₁₅N₃O₂
Molekylvægt: 269.3 g/mol
Smeltepunkt: 178…182 °C
Massefylde: 1.31 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002425
CAS-nummer: 493-52-7
EINECS: 207-776-1
Merck Index: 12,06199

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) slightly soluble
Melting Point 178 - 182 °C
Molar Mass 269.31 g/mol
Bulk Density 300 - 500 kg/m3
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR