F(ab’)2 Anti-IgG Goat Polyclonal Antibody (CF®750)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20920-50UL 20920-250UL
20920-50ULEA 765 DKK
20920-50UL 20920-250UL
F(ab’)2 Anti-IgG Goat Polyclonal Antibody (CF®750)
Antistoffer
Goat anti-mouse IgG (H+L), F(ab’)2 fragment secondary antibody labeled with CF®750 dye.
Suitable for western, immunofluorescence, and immunohistology in FFPE tissues.
Type: Secondary
Antigen: Mouse IgG
Clonality: Polyclonal
Clone:
Conjugation: CF®750
Epitope:
Host: Goat
Isotype: IgG (H+L)
Reactivity: Mouse