Anti-Guinea Pig IgG Donkey Polyclonal Antibody (CF® 583R)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20893-500UL 20893-50UL
20893-500ULEA 1850 DKK
20893-500UL 20893-50UL
Anti-Guinea Pig IgG Donkey Polyclonal Antibody (CF® 583R)
Antistoffer
This is a highly cross-adsorbed donkey anti-guinea pig IgG (H L) secondary antibody labeled with our bright and photostable CF® Dyes. To minimise cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, syrian hamster, horse, human, mouse, rabbit, and sheep serum.

  • Highly cross-adsorbed for specific staining with minimal background
  • Available in 12 bright and photostable CF® Dyes
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues

Conjugates of blue fluorescent dyes like CF®405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Liquid format: Store at −20 °C, protected from light. Product is stable for at least 6 months from date of receipt when stored as recommended. Liquid format antibodies contain 50% glycerol and will not freeze at −20 °C.

Lyophilized format: Store at −20 °C, protected from light. Product is stable for at least 6 months from date of receipt when stored as recommended. Reconstitute antibodies in water using the indicated volumes below:
CF® Dye and biotin conjugates: add 0,5 ml dH₂O
HRP or DNP conjugates: add 1 ml dH₂O
Add the indicated volume of water directly to the vial containing the lyophilised antibody and mix gently to dissolve. Store reconstituted antibody at −20 °C and protect from light. Aliquot to avoid repeated freeze/thaw cycles. Alternatively, an equal volume of glycerol can be mixed with the reconstituted antibody so that it will remain liquid at −20 °C.

Optional: A preservative such as 0,05% sodium azide (final concentration) can be added to CF® Dye and biotin conjugates. Do not add sodium azide to HRP conjugates. Storage of the antibody for more than a day at final working dilution is not recommended.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF583R
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Guinea Pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR