Cyanocobalamin

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ALX460024G001 ALX460024G005
ENZOALX460024G001EA 641 DKK
ENZOALX460024G001 ENZOALX460024G005
Cyanocobalamin
Vitamin B12 (cyanocobalamin)
Formel: C₆₃H₈₈CoN₁₄O₁₄P
Smeltepunkt: 320 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00151092
CAS-nummer: 68-19-9
EINECS: 200-680-0
Merck Index: 12,10152

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR