20-Hydroxyecdysone ≥95% (by 1H-NMR), white powder

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: β-Ecdysone

ALX370012M005 ALX370012M010 ALX370012M050
ENZOALX370012M005EA 776 DKK
ENZOALX370012M005 ENZOALX370012M010 ENZOALX370012M050
20-Hydroxyecdysone ≥95% (by 1H-NMR), white powder
20-Hydroxyecdysone

Ecdysteroid.


Most widely occurring ecdysteroid in both plant and animal species. Controls cell death during metamorphosis of Drosophila melanogaster.


Soluble in water, 100% ethanol, methanol, DMSO and acetic acid.

Formel: C₂₇H₄₄O₇
Opbevaringstemperatur: Fryser
MDL Number: MFCD00036740
CAS-nummer: 5289-74-7

Bestil nu

Specifikationer Testresultater

Purity ≥95% (¹H-NMR)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR