Hypericin (native) ≥98% (by HPLC)
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: 1,3,4,6,8,13-Hexahydroxy-10,11-dimethylphenanthro[1,10,9,8-opqra]perylene-7,14-dione P-conformer
ALX350030M001 ALX350030M005 ALX350030M010
ENZOALX350030M001EA 799 DKK
ENZOALX350030M001 ENZOALX350030M005 ENZOALX350030M010
Hypericin (native) ≥98% (by HPLC)
Hypericin
PKC inhibitor
A potent and selective inhibitor of protein kinase C (IC50=3.4µM). Anti-viral and anti-retroviral actitity. Displays antineoplastic and antitumor actitities. Most activities require light activation. Induces apoptosis in neuroblastoma cells at 1µM or above (IC50=0.1µM at 29 mW/cm2 (tungsten light)). Inhibitition of other kinases (e.g. EGFR-PTK, CKII and MAP kinase). Bright red fluorescence emissino and photostability. Antidepressant.
Isolated from Hypericum perforatum.
Formel: C₃₀H₁₆O₈
Molekylvægt: 504.45 g/mol
MDL Number: MFCD00016683
CAS-nummer: 548-04-9