Curcumin (from Curcuma longa) ≥98% (by HPLC), high purity
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
ALX350028M010 ALX350028M050 ALX350028M250
ENZOALX350028M010EA 386 DKK
ENZOALX350028M010 ENZOALX350028M050 ENZOALX350028M250
Curcumin (from Curcuma longa) ≥98% (by HPLC), high purity
Curcumin
Formel: C₂₁H₂₀O₆
Molekylvægt: 368.39 g/mol
MDL Number: MFCD00008365
CAS-nummer: 458-37-7
EINECS: 207-280-5