bpV(HOpic) ≥90% (by 51V-NMR)
Leverandør: ENZO LIFE SCIENCES
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ALX270206M005
ALX270206M025
ENZOALX270206M005EA
978
DKK
ENZOALX270206M005
ENZOALX270206M025
bpV(HOpic) ≥90% (by 51V-NMR)
bpV(HOpic)
PTP inhibitor, PTEN Inhibitor
Protein phosphotyrosine phosphatase inhibitor.
Formel:
K₂[VO(O₂)₂C₆H₄NO₃] |
CAS-nummer:
722494-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...