GW-9662 ≥98% (by HPLC)
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: GW9662, 2-Chloro-5-nitro-N-phenylbenzamide
BMLGR2340010 BMLGR2340050
ENZOBMLGR2340010EA 866 DKK
ENZOBMLGR2340010 ENZOBMLGR2340050
GW-9662 ≥98% (by HPLC)
GW-9662
PPAR ligand.
Potent, selective and irreversible PPARγ antagonist. Blocks the PPARγ-induced differentiation of monocytes to osteoclasts by >90% at a dose of 0.1 µM. Can be used to antagonise the effects of PPARγ agonists in a variety of cell types.
Soluble in DMSO (50 mg/ml), 100% ethanol (10 mg/ml) or dimethyl formamide. Insoluble in water.
Formel: C₁₃H₉N₂O₃Cl
Molekylvægt: 276.7 g/mol
Smeltepunkt: 155…157 °C
MDL Number: MFCD01215270
CAS-nummer: 22978-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e921714d1c9b480af8862b60e9530751471116d&vid=930ede030b5767df965756e8f6c6b3f9981d185b&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Purity ≥98% (HPLC)