TOFA ≥99% (by TLC)
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BMLEI3210010 BMLEI3210050
ENZOBMLEI3210010EA 665 DKK
ENZOBMLEI3210010 ENZOBMLEI3210050
TOFA ≥99% (by TLC)
TOFA
Acetyl-CoA inhibitor
Hypolipidemic agent. Potent, reversible and competitive inhibitor of acetyl-coenzyme A carboxylase (Ac-CoA carboxylase; ACC), a key enzyme involved in fatty acid biosynthesis. It blocks apoptosis induced by the fatty acid synthase inhibitor C75 (Prod. No. ALX-270-286) and stimulates neurite outgrowth and neuronal differentiation in rat pheochromocytoma cells.
Formel: C₁₉H₃₂O₄
Molekylvægt: 324.461 g/mol
MDL Number: MFCD01726059
CAS-nummer: 54857-86-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6e6fa16b9857340b82df751f5cb9017aa59389f&vid=1d4a03e744c1b07a8f6758c144c1e52543d02907&prid=web&clid=SAPDAM