Rolipram ≥98% (by TLC)

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-(3-(Cyclopentyloxy)-4-methoxyphenyl)pyrrolidin-2-one, 4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-2-pyrrolidinone

BMLPD1750010 BMLPD1750050
ENZOBMLPD1750010EA 632 DKK
ENZOBMLPD1750010 ENZOBMLPD1750050
Rolipram ≥98% (by TLC)
Rolipram

PDE inhibitor


Selective, cell permeable inhibitor of cAMP-specific phosphodiesterase (PDE IV, IC50=1 µM). A rolipram insensitive PDE IV subtype has been described.

Formel: C₁₆H₂₁NO₃
Molekylvægt: 275.35 g/mol
MDL Number: MFCD00270906
CAS-nummer: 61413-54-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=177c24a1c668382ffd0e475a8740725107f7bb82&vid=a98b4b08094c50ed6c52ab3efe656ed449d5c6af&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR