Phytosphingosine ≥98%

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (2S,3S,4R)-2-Aminooctadecane-1,3,4-triol, (2S,3S,4R)-2-Amino-1,3,4-octadecanetriol

BMLSL1500005 BMLSL1500025
ENZOBMLSL1500005EA 665 DKK
ENZOBMLSL1500005 ENZOBMLSL1500025
Phytosphingosine ≥98%
Phytosphingosine

Ultra-pure


An endogenous sphingosine occurring in Saccharomyces cerevisiae. Inhibits FMLP-induced oxidant release by human neutrophils.

Formel: C₁₈H₃₉NO₃
Molekylvægt: 317.51 g/mol
MDL Number: MFCD00079232
CAS-nummer: 554-62-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1f34c340112e09534474ea51457c5230afe77a9&vid=adff813f3b7498f64cbe691600d8cbfd2eba7ce6&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR