Bilobalide ≥98% (by HPLC), white solid

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BMLN1400010 BMLN1400050
ENZOBMLN1400010EA 632 DKK
ENZOBMLN1400010 ENZOBMLN1400050
Bilobalide ≥98% (by HPLC), white solid
Bilobalide

Bilobalide is a terpenoid isolated from Ginkgo biloba. It has been proposed to exert trophic and protective effects on neurons. It increases the sensitivity of mouse pancreatic β cells to glucose and displays protective effects during hypoxia.


Soluble in acetone (clear colourless solution at 25 mg/ml).

Formel: C₁₅H₁₈O₈
Opbevaringstemperatur: Fryser
MDL Number: MFCD00238547
CAS-nummer: 33570-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7407d12c1623d5faa3bd4c1de680030fb10bda7b&vid=cdf8228000427cda59eec34f66d146ab180e01ab&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR