Retinoic acid all trans ≥95%, yellow solid
Leverandør: ENZO LIFE SCIENCESSamlet vurdering
Gennemsntilig vurdering
Synonymer:
all-trans-Retinoic acid, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid, ATRA, Vitamin A acid, Tretinoin
Retinoic acid receptor ligand
Retinoic acid is the active metabolite of vitamin A. It is a potent modulator of growth and differentiation exerting its effects by binding to nuclear retinoic acid receptors (RARs) which directly regulate gene expression. All trans retinoic acid binds to RAR α, β and γ subtypes with Kd values of 0.2, 0.4 and 0.2 nM respectively. RARs function as ligand-dependent transcription factors. Retinoic Acid also promotes the differentiation of mouse embryonic stem cells into adipocytes, neurons and glia in vitro. It guides development of the posterior part of the embryo.
Soluble in DMSO (25 mg/ml) or ethanol (10 mg/ml).
Formel:
C₂₀H₂₈O₂ Molekylvægt: 300.44 g/mol Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00001551 CAS-nummer: 302-79-4 EINECS: 206-129-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...