Anti-Guinea Pig Goat Polyclonal Antibody (CF®583R)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20900-500UL 20900-50UL
20900-500ULEA 1780 DKK
20900-500UL 20900-50UL
Anti-Guinea Pig Goat Polyclonal Antibody (CF®583R)
Antistoffer
Highly cross-adsorbed goat anti-guinea pig IgG (H L) secondary antibody labeled with our superior CF® Dyes.

  • Highly-cross adsorbed for specific staining with minimal background
  • Available in 15 bright and photostable CF® Dyes
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues
  • CF® Dye and biotin conjugates: add 0.5 ml distilled water
  • HRP or DNP conjugates: add 1 ml distilled water

This is a highly cross-adsorbed goat anti-guinea pig IgG (H L) secondary antibody labeled with our bright and photostable CF® Dyes. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, Syrian hamster, horse, human, mouse, rabbit, rat, and sheep serum.

Type: Secondary
Antigen: Guinea pig
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Guinea pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR