Screw Cap GL 40
Leverandør: RIXIUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8-1003-40 U8S1001401800
RIXI8-1003-40EA 11020 DKK
RIXI8-1003-40 RIXIU8S1001401800
Screw Cap GL 40
Låg Flaskelåg
Screw cap made of black PP with ALU laminated PE foam insert for thread GL 40.
  • Height: 14,4 mm
  • Weight: 6 g
The PP screw caps makes it very popular for packaging food and cosmetics, among other things.