WHEATON® PETG, Media, Bottle, Caps
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
W017824 W017828
WHEAW017824EA 0 DKK
WHEAW017824 WHEAW017828
WHEATON® PETG, Media, Bottle, Caps
Låg Flaskelåg
WHEATON® closures are made with medical grade, FDA approved polypropylene and high density polyethylene.
  • Autoclavable, reusable and recyclable
  • Caps are manufactured in a Class 10K clean room environment
  • Caps are supplied non- sterile as well as gamma irradiation sterilised
Tamper evident cap is available for all sizes of WHEATON® PETG and PC media bottles.
Linerless with an integrally-moulded seal ring to ensure a leak proof performance.