Microcentrifuge tubes, 0,5/0,7/1,5/2,0/2,2 ml
Leverandør: Greiner Bio-One
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
616201_4000 623201_4000 667201_10000 716201_500 717201_500 616283_4000
211-2520EA 514 DKK
211-2520 211-2521 211-2522 525-0489 525-0490 525-0488
Microcentrifuge tubes, 0,5/0,7/1,5/2,0/2,2 ml
Rør Mikrocentrifugerør
PP.
  • High chemical and temperature resistance
  • Available for Eppendorf systems (except 211-2522: Vitatron and 211-2524: Roche)
  • Free of detectable DNase, RNase, human DNA, non-pyrogenic