Multipurpose containers
Leverandør: Greiner Bio-One
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
189170_700 201170_400 217111_600 225170_120 229170_48 229180_48 225180_120 201150_400 201151_400 201152_400 201171_400 201172_400 189175_700
GREI189170_700EA 1340 DKK
GREI189170_700 212-3164 212-3176 216-0600 216-0602 216-0603 216-0601 216-0595 GREI201151_400 216-0596 GREI201171_400 216-0597 GREI189175_700
Multipurpose containers
Prøvecontainere Allround kasser
PS, crystal-clear, with snap lid or screw cap.
  • Manufactured under aseptically conditions into a Class II Clean Room environment
  • For single use only
  • Temperature range –20 to +60 °C
Can be used universally for academic and medical purposes.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904