WHEATON® Polypropylene Conical Graduated Centrifuge Tubes
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
380100 380110 380220 380230 380240
WHEA380100EA 3110 DKK
WHEA380100 WHEA380110 WHEA380220 WHEA380230 WHEA380240
WHEATON® Polypropylene Conical Graduated Centrifuge Tubes
Rør Centrifugerør
Ideal for centrifugation and general liquid handling purposes, especially in life science applications, helping eliminating the risk of cross contamination.
  • Manufactured from virgin, medical grade polypropylene
  • Fitted with leakproof, HDPE caps
  • Option of 15 or 50 ml capacity
  • Crisp black printed graduations with large white write on area
  • Supplied sterile by ETO
  • Supplied DNase/RNase-free, non-pyrogenic <0,25 EU/ml
  • Packed in bags