(±)-α-Methoxyphenyleddikesyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: DL-α-Methoxyphenylacetic acid, 2-Methoxy-2-phenylacetic acid, α-Methoxyphenylacetic acid

M0837-25G M0837-5G
TCIAM0837-25GEA 2370 DKK
TCIAM0837-25G TCIAM0837-5G
(±)-α-Methoxyphenyleddikesyre ≥98.0% (by GC, titration analysis)
(±)-α-Methoxyphenyleddikesyre
Formel: C₉H₁₀O₃
Kogepunkt: 137°C
Smeltepunkt: 69…71 °C
MDL Number: MFCD00064217
CAS-nummer: 7021-09-2
EINECS: 230-300-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8e5f9384b331e66753823e0b786121f159fe78d&vid=e8e5f9384b331e66753823e0b786121f159fe78d&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR