Chloramphenicol ≥99%, off-white powder

Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol

0219032191 0219032105 0219032125 0219032180 0219032190
ICNA0219032191EA 7290 DKK
ICNA0219032191 ICNA0219032105 ICNA0219032125 ICNA0219032180 ICNA0219032190
Chloramphenicol ≥99%, off-white powder
Chloramphenicol

Chloramphenicol is a synthetic antibiotic, which was first isolated from strains of Streptomyces venezuelae


It has a broad spectrum of activity against Gram-positive and Gram-negative bacteria.


Mode of Action: Inhibits translation on the 50S ribosomal subunit at the peptidyl transferase step (elongation inhibition). It is bacteriostatic. Mode of Resistance: Acetylation by chloramphenicol acetyltransferase.


Soluble in methanol (50 mg/ml - clear, pale yellow solution), butanol, ethyl acetate, acetone, ethanol, propylene glycol; slightly soluble in water, ether; insoluble in benzene, petroleum ether, vegetable oils.

Formel: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
Molekylvægt: 323.13 g/mol
Kogepunkt: 645
Smeltepunkt: 150
Massefylde: 1.586 g/ml
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00078159
CAS-nummer: 56-75-7
EINECS: 200-287-4
Merck Index: 13,02087

Bestil nu

Specifikationer Testresultater

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Melting point 145 - 155 °C
Optical Rotation (c=5 ethanol) +18° ± 5°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR