5α-Cholestan-3-one ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

L08726.03 L08726.06
L08726.03EA 787 DKK
L08726.03 L08726.06
5α-Cholestan-3-one ≥97%
5α-Cholestan-3-one

Purity: 97% CAS No.: 566-88-1 Molecular Formula: C27H46O Formula Weight: 386.66 Melting Point: 128-132° Beilstein Reference: 2625580

Formel: C₂₇H₄₆O
Smeltepunkt: 128…132 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00065901
CAS-nummer: 566-88-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR