1,3,5-Triiodobenzene ≥95% (by GC), solid
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
T4186-200MG
T4186-1G
TCIAT4186-200MGEA
362
DKK
TCIAT4186-200MG
TCIAT4186-1G
1,3,5-Triiodobenzene ≥95% (by GC), solid
1,3,5-Triiodobenzene
Formel:
C₆H₃I₃ Smeltepunkt: 183 °C Opbevaringstemperatur: Room Temperature |
MDL Number:
MFCD02690052 CAS-nummer: 626-44-8 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity(GC) | min. 95.0% |
confirm to structure | 181.0...185.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...