(4-Nitrophenyl)acetonitril ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Nitrobenzyl cyanide

A14821.18 A14821.30
A14821.18EA 498 DKK
A14821.18 A14821.30
(4-Nitrophenyl)acetonitril ≥98%
(4-Nitrophenyl)acetonitril

Purity: 98% CAS No.: 555-21-5 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Melting Point: 114-117° Boiling Point: 195-197°/12mm Merck Reference: 14,6590 UN No.: 3439 MDL No.: MFCD00007372

Formel: C₈H₆N₂O₂
Molekylvægt: 162.15 g/mol
Kogepunkt: 195…197 °C (12 mmHg)
Smeltepunkt: 114…117 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007372
CAS-nummer: 555-21-5
EINECS: 209-085-0
UN nummer: 3439
ADR: 6.1 III
Merck Index: 12,06688

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR