Clear Polyethylene Cleanroom Bags
Leverandør: KNF CLEANROOM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LB1041014 LB1042030
KNFCLB1041014EA 0 DKK
KNFCLB1041014 KNFCLB1042030
Clear Polyethylene Cleanroom Bags
Poser Prøveposer
Ultraclean polybags can safely store parts and/or materials that require a moderate level (Level II and Level III) of packaging cleanliness.
Polyethylene is an ideal material for general-purpose packaging; has a tensile strength of 2100 psi, and a melting point of 107 °C (225 °F). Approved for cleanroom use in pharmaceuticals, aerospace, semiconductors, electronics, food packaging and many other medical or mechanical applications. Antistatic bags are amine-free; ideal for general-purpose packaging applications requiring antistatic properties.
Certificeringer: Standard polyethylene bagsMore Product Information