Real Time PCR Detection Test Kits, Supreme HLVd (Hop Latent Viroid)
Leverandør: BIOPREMIER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SFS-SP0064 SFS-0064
BIOPSFS-SP0064EA 5710 DKK
BIOPSFS-SP0064 BIOPSFS-0064
Real Time PCR Detection Test Kits, Supreme HLVd (Hop Latent Viroid)
Nukleinsyrereagenser qPCR/RT-qPCR Enzymes and Kits
SUPREME RT-PCR detection kit HLVd is designed for the detection of Hop Latent Viroid (HLVd) in a single PCR reaction. The kit is suitable for use in real time instruments with FAM and Cy5 detection channels.
  • Detection of Hop latent viroid (HLVd) in Cannabis or other plant samples
  • Amplification and detection of HLVd
  • Transcription ensured by the enzyme Reverse transcriptase
  • Real-time PCR with rapid hot-start Taq DNA polymerase
  • Internal control to exclude false-negative results
  • Optimised to handle PCR inhibitors, containing UDG enzyme
  • Higher stability, enzymes in seperate tubes
  • PCR- platforms: runs on all established standard real-time PCR- platforms
This kit HLVd combines a reverse transcriptase with a multiplex qPCR assay to simultaneously detect HLVd and an internal control. The kit is suitable for use in real time instruments with FAM and Cy5 detection channels.
This detection kit HLVd uses Reverse Transcriptase (RT) real-time PCR for the detection of HLVd in a simple, reliable, and rapid procedure. The assay combines a reverse transcriptase with a multiplex qPCR assay based on 5' nuclease real time PCR reactions to amplify a unique genomic sequence of each target. The carefully designed primers and probe ensure the highest sensitivity and specificity. The kit consists of an assay mix for detecting the targets, a positive control, and internal control (IC).To minimise PCR cross-contamination, the kit contains dUTP and uracil-N glycosylase (UDG).
Channel target: FAM
Channel IC: Cy5