3M™ Temflex™ Vinyl Electrical Tape 165
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
165BL5E 165YL5E 165RD5E 165BR5E 165GR5E
MMMA165BL5EEA 2260 DKK
MMMA165BL5E MMMA165YL5E MMMA165RD5E MMMA165BR5E MMMA165GR5E
3M™ Temflex™ Vinyl Electrical Tape 165
Tape
3M™ Temflex™ vinyl electrical tape 165 is a multi-purpose, general use grade, nominal 6-mil (0.152 mm) thick electrical insulating tape composed of an elastic vinyl (PVC) film backing and coated on one side with a non-corrosive, solvent free pressure-sensitive adhesive.
  • Marking of safety areas and equipment and phase coding
  • Indoor and weather protected outdoor operations
  • Wire pulling and fishing
  • Flame retardant and cold resistance rating is required – UL510
  • Industrial, commercial and residential environments