Round Bottom Flasks with Rim
Leverandør: TECHNICAL QUARTZ SOLUTIONS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
75600 75601 75602 75603
TSLA75600EA 2770 DKK
TSLA75600 TSLA75601 TSLA75602 TSLA75603
Round Bottom Flasks with Rim
Kolber Kolber med rund bund
Temperature-resistant round bottom flasks made from transparent quartz glass.
  • Excellent chemical resistance
  • High resistance against thermal shock
  • Made for tasks up to 1050 °C
  • High purity
  • Transparent
The TQS quartz glass round bottom flasks are made for tasks up to 1050 °C (maximum limit 1280 °C). High chemical resistance and purity characterise these products. The durability is dependant on its use and application. Products have a long life for frequent use with temperatures up to 1050 °C.