Poly(styrene-divinylbenzene), microspheres 6.0-10.0 μm avg. part. size, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: SDVB resin, Styrene-divinylbenzene resin, Styrene-divinylbenzene copolymer, Poly(styrene-co-divinylbenzene), Polystyrene crosslinked with divinylbenzene
468312-100G 468312-250G
468312-100GEA 744 DKK
468312-100G 468312-250G
Poly(styrene-divinylbenzene), microspheres 6.0-10.0 μm avg. part. size, Supelco®
Poly(styrene-divinylbenzene)
Formel: [-CH₂CH(C₆H₅)-]x[-CH₂CH[C₆H₄(CHCH₂)]-]y
Molekylvægt: 234.342 g/mol
MDL Number: MFCD00146442
CAS-nummer: 9052-95-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1239c1e425fef012b4dafe365ba84a7e055957c6&vid=85fa8f72de1fcaae21ec5c7ac64ee840d111841a&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63ad89f93519138758b98ef0159a1565c465e715&vid=509fca0f55679a1dcdabc17821b19dd143bab998&prid=web&clid=SAPDAM