Rotors for Centrifuges 5910 Ri and 5920 R
Leverandør: EPPENDORF
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5895120.008 5895121.004 5895130.003 5895135.005 5895150.004 5895200.001 5895201.008 5895175.007
521-2500EA 24900 DKK
521-2500 521-2501 521-2506 521-2507 521-2508 521-2332 521-2333 521-2689
Rotors for Centrifuges 5910 Ri and 5920 R
Centrifuger Rotorer
Eppendorf rotors for centrifuges 5910 Ri and 5920 R - precision in refrigerated large-capacity centrifugation. Tailor your experiments with varied capacities and speeds. Accessorise with versatile buckets and adaptors for efficient sample processing. Ideal for high-throughput applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customise your centrifuge setup
  • Aerosol-tight options available
Experience precision in refrigerated large-capacity centrifugation with Eppendorf rotors designed for centrifuges 5910 Ri and 5920 R. These rotors offer a range of capacities and speeds, providing flexibility for various sample processing needs. Accessorise with versatile buckets and adaptors, enhancing the adaptability of your centrifuge setup. Ideal for high-throughput applications, the Centrifuges 5910 Ri and 5920 R are further optimised with these precision-engineered rotors, ensuring accuracy and reproducibility in every spin. Explore the range to elevate your refrigerated large-capacity centrifugation experience.