Bottles, Wide Neck, Square, Without Screw Cap
Leverandør: RIXIUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11301010032OSV 11301025045OSV 11301050054OSV 11301100054OSV
215-4667EA 697 DKK
215-4667 215-4668 215-4669 215-4670
Bottles, Wide Neck, Square, Without Screw Cap
Flasker Glasflasker
Soda-lime glass, amber.
  • Good chemical and physical properties
  • For use in applications without high thermal loads
DIN thread. Space saving in storage and transport. Stable, non-slip. Amber coloured to protect light-sensitive materials.
Bestillingsoplysninger: Screw caps must be ordered separately.