Phenylacetylchlorid ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B24987.0B
B24987.22
B24987.36
B24987.0BEA
3040
DKK
B24987.0B
B24987.22
B24987.36
Phenylacetylchlorid ≥98%
Phenylacetylchlorid
Purity: 98%
CAS No.: 103-80-0
Molecular Formula: C8H7ClO
Formula Weight: 154.60
Flash Point: 102°(215°F)
Refractive Index: 1.5325
Boiling Point: 225°
Beilstein Reference: 742254
UN No.: 2577
MDL No.: MFCD00000729
Formel:
C₈H₇ClO Molekylvægt: 154.6 g/mol Kogepunkt: 225 °C Smeltepunkt: 91 Massefylde: 1.168 Flammepunkt: 102 °C (215 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000729 CAS-nummer: 103-80-0 EINECS: 203-146-5 UN nummer: 2577 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...