Natriumhydroxid standardised solution 0.01 N in aqueous solution
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Caustic soda
35620.K2 35620.M4
35620.K2EA 455 DKK
35620.K2 35620.M4
Natriumhydroxid standardised solution 0.01 N in aqueous solution
Sodium hydroxide
CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00003548
Formel: NaOH
Molekylvægt: 40 g/mol
Kogepunkt: 1390
Smeltepunkt: 323
Massefylde: 1 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
UN nummer: 1824
ADR: 8 III
Merck Index: 14,08627